Gelite™ Safe DNA Gel Stain *GelRed Alternative, 10000X in DMSO*
Catalog # AATB17711
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- Conf:0,5 mL
- Descrizione:Gelite™ Safe DNA gel stain *GelRed alternative, 10000X in DMSO*
Specifications
About this item
Gelite™ Safe has been developed specifically to be less hazardous than EtBr for staining DNA in agarose and acrylamide gels with much higher sensitivity.
- Stable for 12 months
AAT Bioquest is committed to designing our products to be environment-friendly.
Caution: For research use only.