Amplite® Colorimetric Asparaginase Activity Assay Kit
Catalog # AATB11800
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- Conf:100 Tests
Specifications
About this item
Designed for consistent and dependable results. The Amplite® colorimetric asparaginase activity assay kit provides a simple and straightforward procedure for measuring asparaginase activity in a variety of biological samples.
- Robust protocol
- Absorbance measured at 575 nm
- Six-month shelf life
One unit (U) is the amount of enzyme that catalyses the reaction of 1 µmol of substrate per minute.
Delivery: Shipped with blue ice.
Caution: Research use only.