Amplite® Colorimetric Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) Assay Kit
Codice catalogo AATB11321
Nome del fornitore: AAT Bioquest
New Product
Numero CAS:
Specifiche
- Conf:100 Tests
Specifiche
Informazioni su questo articolo
Designed for consistent and dependable results. The Amplite® colorimetric GAPDH activity assay kit is a sensitive and simple tool for monitoring GAPDH activity based on a coupled enzymatic reaction catalysed by GAPDH.
- Robust protocol
- Absorbance measured at 450 nm
- Six month shelf life
One unit (U) is the amount of enzyme that catalyses the reaction of 1 µmol of substrate per minute.
Delivery: Shipped with blue ice.
Caution: Research use only.