ReadiLink™ Rapid iFluor® 350 Antibody Labelling Kit
Catalog # AATB5700
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Conf:3
- Pk Info:3×1 mg
Specifications
About this item
ReadiLink™ Rapid antibody labelling kits, designed for production scale, provide a convenient and efficient method for labelling large volumes of antibodies with our superior iFluor® dyes.
- Robust protocol
- Consistent results
- Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery: Shipped at RT.
Caution: Research use only.