ContactUsLinkComponent
Amplite® Fluorimetric Asparaginase Activity Assay Kit
Amplite® Fluorimetric Asparaginase Activity Assay Kit
Catalog # AATB11799
Supplier:  AAT Bioquest
New Product
CAS Number:  
Amplite® Fluorimetric Asparaginase Activity Assay Kit
Catalog # AATB11799
Supplier:  AAT Bioquest
New Product
CAS Number:  
Specifications
  • Conf:
    100 Tests
  • Test per kit:
    100 tests
Specifications
About this item
Designed for consistent and dependable results. The Amplite® Fluorimetric Asparaginase Activity Assay Kit provides a simple and straightforward procedure for measuring Asparaginase activity in a variety of biological samples.
  • Ex/Em=540/590 nm
  • Consistent results
  • Six-month shelf life
One unit (U) is the amount of enzyme that catalyzes the reaction of 1 µmol of substrate per minute.
Delivery: Shipped with blue ice.
Caution: Research use only.