Amplite® Colorimetric Hexokinase Assay Kit
Catalog # AATB11319
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- Conf:100 Tests
- Test per kit:100 tests
Specifications
About this item
Designed for consistent and dependable results. The Amplite® Colorimetric Hexokinase Assay Kit is designed to accurately measure the activity of hexokinases from various sources using a simple and quick protocol.
- Robust protocol
- Absorbance measured at 450 nm
- Six month shelf life
This kit utilises a coupled enzyme assay, where glucose is phosphorylated to glucose-6-phosphate and subsequently oxidised to produce NADH.
Delivery: Shipped with blue ice.
Caution: Research use only.