SO Green™ 520AM Singlet Oxygen Sensor *Cell-Permeable*
AATB16062
New Product
:
- Conf:100 µG
- Descrizione:SO Green™ 520AM singlet oxygen sensor *cell-permeable*
SO Green™ 520AM Singlet oxygen sensor has been developed for detecting intracellular singlet oxygen, it is cell-permeant.
- Stable for 24 months
Quality tested in-house.
: For research use only.