ProLite™ 5 to 245 kD Protein Ladder
AATB18015
New Product
:
- Conf:100 µl
- Descrizione:ProLite™ 5-245 kD protein ladder
The ProLite™ 5 to 245 kD protein ladder is a high-quality three-colour protein standard consisting of 13 prestained proteins.
- Stable for 12 months
Quality tested in-house.
: For research use only.