iFluor® 532-Wheat Germ Agglutinin (WGA) Conjugate
Catalog # AATB25532
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Conf:1 mg
- Descrizione:iFluor® 532-Wheat germ agglutinin (WGA) conjugate
Specifications
About this item
iFluor® 532 WGA conjugate, like Alexa Fluor® 532 WGA conjugate, retains its ability to bind to sialic acid and N-acetylglucosaminyl residues, enhancing its utility in fluorescence imaging and analysis for various scientific investigations.
- Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
Caution: For research use only.