Digital Solutions
Applications
Support
About
Inserisci ordini
Stato dell'ordine
Il tuo profilo
Carrello
Switzerland
Inserisci ordini
Stato dell'ordine
Il tuo profilo
Carrello
ContactUsLinkComponent
Pagina principale
XFD568 NHS Ester
XFD568 NHS Ester [Same Structure to Alexa Fluor™ 568 NHS ester]
XFD568 NHS Ester [Same Structure to Alexa Fluor™ 568 NHS ester]
XFD568 NHS Ester [Same Structure to Alexa Fluor™ 568 NHS ester]
Catalog # AATB71907
Supplier:
AAT Bioquest
New Product
CAS Number:
Expand image
XFD568 NHS Ester [Same Structure to Alexa Fluor™ 568 NHS ester]
Catalog # AATB71907
Supplier:
AAT Bioquest
New Product
CAS Number:
Specifications
Conf:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
:
791.8 g/mol
CAS Number:
247145-38-6
Product Details