Orders LinkContactUsLinkComponent
OG488 DHPE
OG488 DHPE
1 mg
Codice catalogo AATB23302
Nome del fornitore:   AAT Bioquest
OG488 DHPE
Codice catalogo AATB23302
Nome del fornitore:  AAT Bioquest
Numero CAS:  
OG488 DHPE
1 mg
Numero CAS:  
Price
SFr. 139.00
Log in per disponibilità
1 * 1 mg
Product Details & Documents
OG488 DHPE is a phospholipid that is labeled on the head group with the bright, green-fluorescent fluorinated fluorescein dye (also called Oregon Green® 488, Oregon Green® is a trademark of Invitrogen).
  • Stable for 12 months
Quality tested in-house
Attenzione: For Research Use Only
  • Molecular Weight:1086.24 g/mol
Specifiche
Specifiche
  • Conf:
    1 mg
Specifiche