Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

2’,3’-cGAMP azide

100 µG
Katalognr. AATB20332
Anbieter:   AAT Bioquest
2’,3’-cGAMP azide
Katalognr. AATB20332
Anbieter:  AAT Bioquest
CAS Nummer:  
2’,3’-cGAMP azide

100 µG

CAS Nummer:  
Preis
SFr. 1’340.00
Einloggen für Verfügbarkeit
1 * 100 µG
Product Details & Documents

2’,3’-cGAMP azide is an excellent reagent to conjugate 2’,3’-cGAMP to alkyne-modified biological targets, such as proteins, peptides, and nucleic acids.

  • Stable for 12 months

Quality tested in-house

 : For Research Use Only

  • Molekulargewicht:1102.05 g/mol
Technische Daten

Technische Daten

  • Conf:
    100 µG

Technische Daten