CytoCite™ Sample Tubes
500 μl tubes, CytoCite™ sample tubes
Codice catalogo AATBCCT100
Nome del fornitore: AAT Bioquest
Vai a
Product Details & Documents
Cytocite™ sample tubes are 500 μl thin-walled polypropylene tubes validated for use with Cytocite or Qubit Fluorometer.
- Storage at room temperature
Optimised for easy use with both common fluorometer instruments.
Attenzione: Research use only.
Specifiche
- Capacità:500 μl tubes
- Descrizione:CytoCite™ sample tubes