PBS Tablet (Phosphate Buffered Saline Tablet)
Codice catalogo AATB10010
Nome del fornitore: AAT BIOQUEST
New Product
Numero CAS:
Specifiche
- Conf:10 Tab.
- Descrizione:Phosphate buffered saline tablet
Specifiche
Informazioni su questo articolo
Each PBS tablet prepares 100 mL of a 1X PBS solution containing 137 mM sodium chloride, 10 mM phosphate buffer, and 2.7 mM potassium chloride (pH 7.4).
- Designed for ease of use
- Saves time
PBS (phosphate buffered saline) is an isotonic buffer that closely mimics the human body's pH, osmolarity, and ion concentrations.
Caution: For research use only.