Order Entry
Switzerland
ContactUsLinkComponent
 
Mouse EDG3/S1P3 ELISA Kit
Catalog # ANTIA77275-96
Supplier:  Antibodies.com
New Product
CAS Number:  
undefined
Mouse EDG3/S1P3 ELISA Kit
Catalog # ANTIA77275-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Specifications

  • Conf:
    96 Tests
  • Durata del test:
    Multiple steps
  • Tipo di test:
    Sandwich
  • Formato:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    EDG3/S1P3
  • Descrizione:
    Mouse EDG3/S1P3 ELISA kit
  • Tiipo del campione:
    Serum, plasma, tissue homogenates and other biological fluids
  • Reattività incrociata:
    Mouse EDG3/S1P3 ELISA kit exhibits high specificity and excellent specificity for the detection of mouse EDG3/S1P3. No significant cross-reactivity or interference between EDG3/S1P3 and analogues was observed.
  • Metodo di rilevamento:
    Colorimetric
  • Tempo ai risultati:
    4 h 30 min
  • Principio del dosaggio:
    Quantitative
  • Durata a magazzino:
    Store for 6 months at 4 °C
  • Range di rilevamento:
    62,5 - 4000 pg/ml
  • Temperatura di conservazione:
    4 °C
  • Volume del campione:
    100 μl
  • Sensibilità:
    37,5 pg/ml
  • Stato regolamentazione:
    RUO

Specifications

About this item

Mouse EDG3/S1P3 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse EDG3/S1P3 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 62,5 to 4000 pg/ml
  • Sensitivity: 37,5 pg/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse EDG3 / S1P3 employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse EDG3 / S1P3 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for EDG3 / S1P3 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the EDG3 / S1P3 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-EDG3 / S1P3 Antibody, which binds the captured EDG3 / S1P3 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of EDG3 / S1P3 captured in each well. The concentration of EDG3 / S1P3 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.