- Conf:96 Tests
- Durata del test:One step
- Tipo di test:Sandwich
- Formato:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:HAS1
- Descrizione:Mouse HAS1 ELISA kit
- Tiipo del campione:Serum, plasma or other biological fluids
- Metodo di rilevamento:Colorimetric
- Tempo ai risultati:1 h 30 min
- Durata a magazzino:Store for 6 months at 4 °C
- Range di rilevamento:30 - 480 ng/L
- Temperatura di conservazione:4 °C
- Volume del campione:40 μl
- Sensibilità:1,53 ng/L
- Stato regolamentazione:RUO
Mouse HAS1 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse HAS1 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection Range: 30 to 480 ng/L
- Sensitivity: 1,53 ng/L
- Sample Type: Serum, plasma or other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Mouse HAS1 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse HAS1 in serum, plasma or other biological fluids. An antibody specific for HAS1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the HAS1 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-HAS1 antibody, which also binds the HAS1 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-HAS1 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-HAS1 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of HAS1 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of HAS1 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.