Specifiche
- Conf:96 Tests
- Durata del test:Multiple steps
- Tipo di test:Sandwich
- Formato:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:PRIM2
- Descrizione:Rat PRIM2 ELISA kit
- Tiipo del campione:Serum, plasma, tissue homogenates and other biological fluids
- Reattività incrociata:Rat PRIM2 ELISA kit exhibits high specificity and excellent specificity for the detection of rat PRIM2. No significant cross-reactivity or interference between PRIM2 and analogues was observed.
- Metodo di rilevamento:Colorimetric
- Tempo ai risultati:4 h 30 min
- Principio del dosaggio:Quantitative
- Durata a magazzino:Store for 6 months at 4 °C
- Range di rilevamento:0,156 - 10 ng/ml
- Temperatura di conservazione:4 °C
- Volume del campione:100 μl
- Sensibilità:0,094 ng/ml
- Stato regolamentazione:RUO
Specifiche
Informazioni su questo articolo
Rat PRIM2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat PRIM2 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 0,156 - 10 ng/ml
- Sensitivity: 0,094 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Rat PRIM2 ELISA kit (A79660) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat PRIM2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PRIM2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PRIM2 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PRIM2 Antibody, which binds the captured PRIM2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PRIM2 captured in each well. The concentration of PRIM2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.