MP650 Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Catalog # AATB67019
Anbieter: AAT Bioquest
New Product
Technische Daten
- Colore del telaio:bianco
- N. di pozzetti:96
- Profilo:Low
- Bordo:Semi-skirted
- Colore del pozzetto:bianco
- Descrizione:MP650 Dye qPCR calibration plate
- Con bar code:Yes
- Conf:1
Technische Daten
Über diesen Artikel
MP650 is designed to have identical spectra to those of Mustang purple.
- The qPCR calibration plate might significantly improve qPCR results with multiplexing by more accurately representing fluorescent spectra used in your real-time experiments
- This calibration plate is ready to use without any additional preparation steps required
For most qPCR instruments, the necessary calibrations should be run at least every six months.
Caution: Avoid freeze-thawing plates. It is not recommended to use the plates more than 3 times. Plates can be stored for up to 12 months. Freeze (<−15 °C), and minimise light exposure.