HygroSoft Software
HygroSoft software
Codice catalogo ROTRHS-AWCODE
Nome del fornitore: ROTRONIC INSTRUMENTS
Vai a
Product Details & Documents
The HygroSoft software is compatible with all Rotronic products with USB, UART or ethernet interfaces and runs on all WINDOWS 10 operating systems.
- Live display of current measured values
- Configuration of instruments
- Probe calibration and adjustment
- AwQuick function
- Measurement within 4 to 5 minutes
- Viewing of measured values is very easy and user-friendly
- The data is presented in both tabular and graphical formats
The HygroSoft software can be used to adjust the settings of Rotronic instruments and probes. Depending on the instrument and probe, the following functions and settings can be changed General device settings, Change the system units: metric /imperial, AwE and AwQuick, Aw mode settings.
Specifiche
- Descrizione:HygroSoft software
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..