Pimecrolimus
100 mg
Codice catalogo 1539100.
Nome del fornitore: USP
Vai a
Product Details & Documents
Sinonimi: (3S,4R,5S,8R,9E,12S,14S,15R,16S,18R,19R,26aS)-3-[(1E)-2-[(1R,3R,4S)-4-Chloro-3-methoxycyclohexyl]-1-methylethenyl]-8-ethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-14,16-dimethoxy-4,10,12,18-tetramethyl-15,19-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclotricosine-1,7,20,21(4H,23H)-tetrone
- Numero CAS:137071-32-0
- Numero MDL:MFCD00901792
Specifiche
Specifiche
- Conf:100 mg
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



