Supelco® Discovery® BIO C5 Reversed-Phase Solution to Protein and Peptide Separation Challenges
: Merck
Discovery® BIO Wide Pore reversed-phase columns satisfy the need for efficiency, selectivity, scalability, and reproducibility for RP HPLC analyses of proteins, peptides, and small biomolecules.
- Enhanced stability and lifetime compared to conventional C4 phases
- Better protein and peptide resolution compared to leading RP-HPLC FPP columns
- High efficiency for peptide mapping
- Low-bleed, inert surface chemistry make them ideal for LC-MS applications
- Reliable reproducibility run -to-run, column-to-column, batch-to-batch
- Separation are completely scalable from analytical to preparative dimensions
- Surface coverage: 4,5 µmol/m²
Discovery® BIO Wide Pore HPLC columns are packed with C5 ligands bonded to 3, 5, or 10 µm spherical, high purity silica particles containing 300 Å pores.
The low-bleed character and excellent peak shape without TFA in the mobile phase makes these columns ideal for proteomics and other LC-MS applications and preparative purification.
: Column test certificate available inside the box.
- Catalog No:
- 567280-U
- 567231-U
- 567230-U
- 567278-U
- 568430-U
- 568423-U
- 568422-U
- 568421-U
- 568472-U
- 568402-U
- 65511-U
- 567234-U
- 567281-U
- 567232-U
- 568420-U
- 567229-U
- 567228-U
- 567227-U
- 567226-U
- Applicazione:
- Hydrophobic peptides/proteins
- Hydrophobic peptides/proteins
- Hydrophobic peptides/proteins
- Hydrophobic peptides/proteins
- Hydrophobic peptides/proteins
- Hydrophobic peptides/proteins
- Hydrophobic peptides/proteins
- Hydrophobic peptides/proteins
- Hydrophobic peptides/proteins
- Hydrophobic peptides/proteins
- Hydrophobic peptides/proteins
- Hydrophobic peptides/proteins
- Hydrophobic peptides/proteins
- Hydrophobic peptides/proteins
- Hydrophobic peptides/proteins
- Hydrophobic peptides/proteins
- Hydrophobic peptides/proteins
- Hydrophobic peptides/proteins
- Hydrophobic peptides/proteins
- Carico di carbonio:
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- 3,5%
- Con endcapping:
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Sì
- Pressione max.:
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- 400 bar
- Range di pH:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Area superficiale:
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- Limite di temperatura:
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- 0...70 °C
- Condizioni di spedizione:
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
- Acetonitril/water
Frequently Bought Together