Laboratory Chair, with Rollers, Isitec 2
Swivel chair, black, foam, with permanent contact mechanism and seat angle adjustment, 430 - 600 mm, Castors
Codice catalogo BIMO9608-2000SO272
Nome del fornitore: bimos
Laboratory Chair, with Rollers, Isitec 2
Codice catalogo BIMO9608-2000SO272
Nome del fornitore: bimos
Vai a
Product Details & Documents
Large, ergonomically designed backrest to take the strain off the spine and muscles with integrated lumbar support.
- The seat and backrest are made from sealed SoftTouch integral foam
- This material can be disinfected and is easy to clean
- Despite Isitec Laboratorys minimalist approach it is still very well equipped making it an attractive budget solution for laboratory applications
- Instead of the sturdy plastic 5 star base it can also be supplied with a steel 5 star base as an option
- The colour of the frame is black
Functions:
Seat height adjustment
Backrest height adjustment
Contact backrest with seat tilt adjustment
Specifiche
- Descrizione:Swivel chair, black, foam, with permanent contact mechanism and seat angle adjustment
- Altezza:430 - 600 mm
- Seq:01
- Base:Castors
- Cat. No.:BIMO9608-2000SO272
- Colore/finitura telaio:Black
- Materiale retro:SoftTouch integral foam
- Modello:Isitec 2
- Materiale del telaio:Steel