ContactUsLinkComponent
Ethyl(2E)-2-cyano-3-(1H-indolyl-3-yl)acrylate 99%, Sigma-Aldrich®
Ethyl(2E)-2-cyano-3-(1H-indolyl-3-yl)acrylate 99%, Sigma-Aldrich®
  E18905-1G
 :  
undefined
Ethyl(2E)-2-cyano-3-(1H-indolyl-3-yl)acrylate 99%, Sigma-Aldrich®
  E18905-1G
 :  
  • Conf:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:
 : Ethyl trans-α-cyano-3-indoleacrylate
 :
 :3077
 :GHS07
 :164...166 °C (dec., lit.)
 :Room Temperature
 :9,III
 :240.26 g/mol
 :62309-96-0
 :C₁₄H₁₂N₂O₂
 :MFCD00022742