Orders LinkContactUsLinkComponent
MiniCorer Sampler
MiniCorer Sampler
Supplier:   Sampling Systems
MiniCorer Sampler
Supplier:  Sampling Systems
Certificates
MiniCorer Sampler
MiniCorer Sampler
3 Options Available Below
Product Details & Documents
The MiniCorer is designed for sampling cheese, wax, butter, etc.
  • Available in multipe lengths
  • Made from chemically resistant 316L stainless steel
  • Available from stock
  • Fast delivery
The MiniCorer is an ideal sampler for sampling cheese, wax, butter, and other similar substances. Available in varying lengths and made from chemically resistant 316L stainless steel, the sampler offers a convenient, quick, and easy way to take core samples.
Product Family Options
Product Information
  • Descrizione
    Materiale
    Lunghezza
    Ø
    Availability
    Price
  • MiniCorer sampler
    Acciaio inossidabile 316L
    190 mm
    18 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 87.50
    SFr. 87.50
    1 * 1 ST
       SFr. 87.50
  • MiniCorer sampler
    Acciaio inossidabile 316L
    235 mm
    18 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 103.00
    SFr. 103.00
    1 * 1 ST
       SFr. 103.00
  • MiniCorer sampler
    Acciaio inossidabile 316L
    290 mm
    18 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 116.00
    SFr. 116.00
    1 * 1 ST
       SFr. 116.00