Deep-Freeze Removable Tapes
Informazioni su questo articolo
Removable lab tape for deep-freeze storage of cryo containers, including freezer boxes, tubes, vials, plastic bags, and other items intended for long-term storage in laboratory freezers and dry ice transportation. Writable using permanent cryo markers.
- Temperature range: −80 to 100 °C (−112 to 212 °F)
- Resists long-term storage in deep-freeze conditions, including multiple freeze-thaw cycles
- Glove-friendly and removable; won't leave behind any adhesive residue
- Available in a range of colors for easy color coding
Deep-freeze removable tape for sealing or identifying cryo and freezer boxes, tubes, vials, plastic bags, and other items intended for long-term storage in laboratory freezers (−20 °C, −40 °C, −80 °C) and for dry ice transportation. Writable with permanent cryo markers, glove-friendly, withstands dry ice, multiple freeze-thaw cycles, gamma irradiation, and other harsh laboratory environments. The tape should be applied at room temperature before storage in freezers. Not recommended for use in liquid nitrogen.
36 varianti
Product Details & Documents
Removable lab tape for deep-freeze storage of cryo containers, including freezer boxes, tubes, vials, plastic bags, and other items intended for long-term storage in laboratory freezers and dry ice transportation. Writable using permanent cryo markers.
- Temperature range: −80 to 100 °C (−112 to 212 °F)
- Resists long-term storage in deep-freeze conditions, including multiple freeze-thaw cycles
- Glove-friendly and removable; won't leave behind any adhesive residue
- Available in a range of colors for easy color coding
Deep-freeze removable tape for sealing or identifying cryo and freezer boxes, tubes, vials, plastic bags, and other items intended for long-term storage in laboratory freezers (−20 °C, −40 °C, −80 °C) and for dry ice transportation. Writable with permanent cryo markers, glove-friendly, withstands dry ice, multiple freeze-thaw cycles, gamma irradiation, and other harsh laboratory environments. The tape should be applied at room temperature before storage in freezers. Not recommended for use in liquid nitrogen.
Product Family Options
Informazioni sul prodotto
- ColoreDimensioniDisponibilitàPrice
- GAINTRM-13C1-50BLDeep-freeze removable tape, core size: 1"Deep-freeze removable tape, core size: 1"blu12,7 mm×15 m (0,5"×50')Log in per disponibilità1 * 1 ROUSFr. 31.60
- GAINTRM-13C1-50GADeep-freeze removable tape, core size: 1"Deep-freeze removable tape, core size: 1"verde12,7 mm×15 m (0,5"×50')Log in per disponibilità1 * 1 ROUSFr. 31.60
- GAINTRM-13C1-50ORDeep-freeze removable tape, core size: 1"Deep-freeze removable tape, core size: 1"arancione12,7 mm×15 m (0,5"×50')Log in per disponibilità1 * 1 ROUSFr. 31.60
- GAINTRM-13C1-50REDeep-freeze removable tape, core size: 1"Deep-freeze removable tape, core size: 1"rosso12,7 mm×15 m (0,5"×50')Log in per disponibilità1 * 1 ROUSFr. 31.60
- GAINTRM-13C1-50WHDeep-freeze removable tape, core size: 1"Deep-freeze removable tape, core size: 1"bianco12,7 mm×15 m (0,5"×50')Log in per disponibilità1 * 1 ROUSFr. 31.60
- GAINTRM-13C1-50YEDeep-freeze removable tape, core size: 1"Deep-freeze removable tape, core size: 1"giallo12,7 mm×15 m (0,5"×50')Log in per disponibilità1 * 1 ROUSFr. 31.60
- GAINTRM-13C3-100BLDeep-freeze removable tape, core size: 3"Deep-freeze removable tape, core size: 3"blu12,7 mm×30 m (0,5"×100')Log in per disponibilità1 * 1 ROUSFr. 63.10
- GAINTRM-13C3-100GADeep-freeze removable tape, core size: 3"Deep-freeze removable tape, core size: 3"verde12,7 mm×30 m (0,5"×100')Log in per disponibilità1 * 1 ROUSFr. 63.10
- GAINTRM-13C3-100ORDeep-freeze removable tape, core size: 3"Deep-freeze removable tape, core size: 3"arancione12,7 mm×30 m (0,5"×100')Log in per disponibilità1 * 1 ROUSFr. 63.10
- GAINTRM-13C3-100REDeep-freeze removable tape, core size: 3"Deep-freeze removable tape, core size: 3"rosso12,7 mm×30 m (0,5"×100')Log in per disponibilità1 * 1 ROUSFr. 63.10
- GAINTRM-13C3-100WHDeep-freeze removable tape, core size: 3"Deep-freeze removable tape, core size: 3"bianco12,7 mm×30 m (0,5"×100')Log in per disponibilità1 * 1 ROUSFr. 63.10
- GAINTRM-13C3-100YEDeep-freeze removable tape, core size: 3"Deep-freeze removable tape, core size: 3"giallo12,7 mm×30 m (0,5"×100')Log in per disponibilità1 * 1 ROUSFr. 63.10
- GAINTRM19SBC1-50BLDeep-freeze removable tape, core size: 1"Deep-freeze removable tape, core size: 1"blu19 mm×15 m (0,75"×50')Log in per disponibilità1 * 1 ROUSFr. 36.80
- GAINTRM19SBC1-50GADeep-freeze removable tape, core size: 1"Deep-freeze removable tape, core size: 1"verde19 mm×15 m (0,75"×50')Log in per disponibilità1 * 1 ROUSFr. 36.80
- GAINTRM19SBC1-50ORDeep-freeze removable tape, core size: 1"Deep-freeze removable tape, core size: 1"arancione19 mm×15 m (0,75"×50')Log in per disponibilità1 * 1 ROUSFr. 36.80
- GAINTRM19SBC1-50REDeep-freeze removable tape, core size: 1"Deep-freeze removable tape, core size: 1"rosso19 mm×15 m (0,75"×50')Log in per disponibilità1 * 1 ROUSFr. 36.80
- GAINTRM19SBC1-50WHDeep-freeze removable tape, core size: 1"Deep-freeze removable tape, core size: 1"bianco19 mm×15 m (0,75"×50')Log in per disponibilità1 * 1 ROUSFr. 36.80
- GAINTRM19SBC1-50YEDeep-freeze removable tape, core size: 1"Deep-freeze removable tape, core size: 1"giallo19 mm×15 m (0,75"×50')Log in per disponibilità1 * 1 ROUSFr. 36.80
- GAINTRM19SBC3100BLDeep-freeze removable tape, core size: 3"Deep-freeze removable tape, core size: 3"blu19 mm×30 m (0,75"×100')Log in per disponibilità1 * 1 ROUSFr. 73.40
- GAINTRM19SBC3100GADeep-freeze removable tape, core size: 3"Deep-freeze removable tape, core size: 3"verde19 mm×30 m (0,75"×100')Log in per disponibilità1 * 1 ROUSFr. 73.40
- GAINTRM19SBC3100ORDeep-freeze removable tape, core size: 3"Deep-freeze removable tape, core size: 3"arancione19 mm×30 m (0,75"×100')Log in per disponibilità1 * 1 ROUSFr. 73.40
- GAINTRM19SBC3100REDeep-freeze removable tape, core size: 3"Deep-freeze removable tape, core size: 3"rosso19 mm×30 m (0,75"×100')Log in per disponibilità1 * 1 ROUSFr. 73.40
- GAINTRM19SBC3100WHDeep-freeze removable tape, core size: 3"Deep-freeze removable tape, core size: 3"bianco19 mm×30 m (0,75"×100')Log in per disponibilità1 * 1 ROUSFr. 73.40
- GAINTRM19SBC3100YEDeep-freeze removable tape, core size: 3"Deep-freeze removable tape, core size: 3"giallo19 mm×30 m (0,75"×100')Log in per disponibilità1 * 1 ROUSFr. 73.40
- GAINTRM25SBC1-50BLDeep-freeze removable tape, core size: 1"Deep-freeze removable tape, core size: 1"blu25,4 mm×15 m (1"×50')Log in per disponibilità1 * 1 ROUSFr. 42.90
- GAINTRM25SBC1-50GADeep-freeze removable tape, core size: 1"Deep-freeze removable tape, core size: 1"verde25,4 mm×15 m (1"×50')Log in per disponibilità1 * 1 ROUSFr. 42.90
- GAINTRM25SBC1-50ORDeep-freeze removable tape, core size: 1"Deep-freeze removable tape, core size: 1"arancione25,4 mm×15 m (1"×50')Log in per disponibilità1 * 1 ROUSFr. 42.90
- GAINTRM25SBC1-50REDeep-freeze removable tape, core size: 1"Deep-freeze removable tape, core size: 1"rosso25,4 mm×15 m (1"×50')Log in per disponibilità1 * 1 ROUSFr. 42.90
- GAINTRM25SBC1-50WHDeep-freeze removable tape, core size: 1"Deep-freeze removable tape, core size: 1"bianco25,4 mm×15 m (1"×50')Log in per disponibilità1 * 1 ROUSFr. 42.90
- GAINTRM25SBC1-50YEDeep-freeze removable tape, core size: 1"Deep-freeze removable tape, core size: 1"giallo25,4 mm×15 m (1"×50')Log in per disponibilità1 * 1 ROUSFr. 42.90
- GAINTRM25SBC3100BLDeep-freeze removable tape, core size: 3"Deep-freeze removable tape, core size: 3"blu25,4 mm×30 m (1"×100')Log in per disponibilità1 * 1 ROUSFr. 85.50
- GAINTRM25SBC3100GADeep-freeze removable tape, core size: 3"Deep-freeze removable tape, core size: 3"verde25,4 mm×30 m (1"×100')Log in per disponibilità1 * 1 ROUSFr. 85.50
- GAINTRM25SBC3100ORDeep-freeze removable tape, core size: 3"Deep-freeze removable tape, core size: 3"arancione25,4 mm×30 m (1"×100')Log in per disponibilità1 * 1 ROUSFr. 85.50
- GAINTRM25SBC3100REDeep-freeze removable tape, core size: 3"Deep-freeze removable tape, core size: 3"rosso25,4 mm×30 m (1"×100')Log in per disponibilità1 * 1 ROUSFr. 85.50
- GAINTRM25SBC3100WHDeep-freeze removable tape, core size: 3"Deep-freeze removable tape, core size: 3"bianco25,4 mm×30 m (1"×100')Log in per disponibilità1 * 1 ROUSFr. 85.50
- GAINTRM25SBC3100YEDeep-freeze removable tape, core size: 3"Deep-freeze removable tape, core size: 3"giallo25,4 mm×30 m (1"×100')Log in per disponibilità1 * 1 ROUSFr. 85.50