GenSwab™ for Collection and Preservation of Biological Samples
Catalog # BINZGS2CGSW
Supplier: Ahlstrom-Munksjö
Specifications
- Sterilità:Sterili
- Descrizione:GenSwab™ with integrated specimen collection card
- Materiale:Foam
- Conf:100
Specifications
About this item
GenSwab™ specifically designed for easy collection and long-term preservation of buccal samples to achieve high recovery of DNA and high-quality genetic profiles.
- Environmentally friendly paper-based design
- Automated design compatible with automated DBS equipment
- Complete protection of samples during transport
GenSwab™ allows easy collection and long-term preservation of buccal samples at ambient temperature, safeguarding their quality over time for downstream processing and improving sample traceability with multiple barcodes.
Certifications: The device is manufactured in compliance with the ISO 18385:2016 standard.