ImmunoCult™-ACF Dendritic Cell Differentiation Supplement
N° de catalogue STMC10988
Fournisseur: STEMCELL Technologies
Numéro CAS:
Spécifications
- Conf:1 KIT
- Descrizione:ImmunoCult™ ACF dendritic cell differentiation supplement
Spécifications
À propos de cet article
Supplement for the differentiation of human monocytes into dendritic cells.
Generate immature dendritic cells (DCs) from human monocytes using animal component-free (ACF) ImmunoCult™-ACF Dendritic Cell Differentiation Supplement. For your convenience, the medium is available either individually or as part of ImmunoCult™ Dendritic Cell Culture Kit. Use the ImmunoCult™-ACF Dendritic Cell Differentiation Supplement in combination with ImmunoCult™-ACF Dendritic Cell Medium to generate immature DCs in vitro.
To support the maturation of immature DCs to mature DCs, combine with the ImmunoCult™ Dendritic Cell Maturation Supplement.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').