Assistent® Brush with Nylon Bristles
Nylon, 8 mm, Acciaio inossidabile, 250 mm
Catalog # HECH91120000258
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Brush with nylon bristles for use with laboratory glassware.
- Black bonded cap
- Stainless steel handle
Specifications
- Materiale setola:Nylon
- Testa della spazzola Ø:8 mm
- Handle material:Acciaio inossidabile
- Lunghezza totale:250 mm