Anti-ELMO2 Rabbit Polyclonal Antibody
Catalog # PRSI55-631
Anbieter: ProSci Inc.
New Product
Technische Daten
- Tipo anticorpo:Primary
- Tipo di anticorpi:Engulfment and cell motility protein 2
- Simbolo dell'antigene:ELMO2
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- Citometria a flusso:Yes
- Ospite:Rabbit
- ImmunoChimica:Yes
- Reattività:Human
- Western blot:Yes
- Epitopo:Amino Acid Range = 323-350
- Forma:Liquid
- ID gene:Q96JJ3
- Sinomini antigene:ELMO2
- Modifica:None
- Tampone di conservazione:Supplied in PBS with 0,09% (W/V) sodium azide
- Peso molecolare:83 kDa
- Temperatura di conservazione:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentrazione:0,39 mg/ml
- Temperatura di spedizione:4 °C
- Immunogeno:This ELMO2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 323-350 amino acids from the Central region of human ELMO2.
- Purificazione:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Taglia:400 µl
- Conf:400 µl
Technische Daten
Über diesen Artikel
The protein encoded by this gene interacts with the dedicator of cyto-kinesis 1 protein. Similarity to a C. elegans protein suggests that this protein may function in phagocytosis of apoptotic cells and in cell migration. Alternative splicing results in multiple transcript variants encoding the same protein.
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: ELMO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human