Anti-ACTC1 Rabbit Polyclonal Antibody
Codice catalogo PRSI58-098
Nome del fornitore: ProSci Inc.
Specifiche
- Tipo anticorpo:Primary
- Tipo di anticorpi:Actin, alpha cardiac muscle 1
- Simbolo dell'antigene:ACTC1
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- Ospite:Rabbit
- ImmunoChimica:Yes
- ImmunoFluorescenza:Yes
- Western blot:Yes
- Epitopo:38 - 67 aa
- Forma:Liquid
- ID gene:70
- Modifica:None
- Peso molecolare:42 kDa
- Temperatura di conservazione:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentrazione:2 mg/ml
- Temperatura di spedizione:4 °C
- Immunogeno:This Actin (ACTB/ACTC) antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 38-67 amino acids from the N-terminal region of human Actin (ACTB/ACTC)
- Purificazione:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Conf:400 µl
Specifiche
Informazioni su questo articolo
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50. For IF starting dilution is: 1:10~50.
Type: Primary
Antigen: ACTC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 38-67
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse