Anti-SLC2A2 Rabbit Polyclonal Antibody
PRSI58-080
- Tipo anticorpo:Primary
- Tipo di anticorpi:Solute carrier family 2, facilitated glucose transporter member 2
- Simbolo dell'antigene:SLC2A2
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- Citometria a flusso:Yes
- Ospite:Rabbit
- Reattività:Human
- Western blot:Yes
- Epitopo:31 - 60 aa
- Forma:Liquid
- ID gene:6514
- Modifica:None
- Peso molecolare:57 kDa
- Temperatura di conservazione:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentrazione:2 mg/ml
- Temperatura di spedizione:4 °C
- Immunogeno:This GLUT2 (SLC2A2) antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 31-60 amino acids from the N-terminal region of human GLUT2 (SLC2A2)
- Purificazione:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Conf:400 µl
For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: SLC2A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 31-60
Host: Rabbit
Isotype: Ig
Reactivity: Human