Anti-PCSK2 Rabbit Polyclonal Antibody
Codice catalogo PRSI63-050
Nome del fornitore: ProSci Inc.
Specifiche
- Tipo anticorpo:Primary
- Tipo di anticorpi:Neuroendocrine convertase 2
- Simbolo dell'antigene:PCSK2
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- Citometria a flusso:Yes
- Ospite:Rabbit
- ImmunoChimica:Yes
- ImmunoFluorescenza:Yes
- Reattività:Human,Mouse
- Western blot:Yes
- Epitopo:87-116 aa
- Forma:Liquid
- ID gene:5126
- Modifica:None
- Peso molecolare:71 kDa
- Temperatura di conservazione:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentrazione:2 mg/ml
- Temperatura di spedizione:4 °C
- Immunogeno:This PCSK2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 87-116 amino acids from the N-terminal region of human PCSK2.
- Purificazione:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Conf:400 µl
Specifiche
Informazioni su questo articolo
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50. For IF starting dilution is: 1:10~50. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: PCSK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 87-116
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse