Orders LinkContactUsLinkComponent
Eppendorf® Microplates
Eppendorf® Microplates
   Opzioni sostenibili disponibili
Nome del fornitore:   EPPENDORF
Eppendorf® Microplates
Nome del fornitore:  EPPENDORF
   Opzioni sostenibili disponibili
Certificates
Eppendorf® Microplates
13 varianti
Product Details & Documents
Eppendorf microplates provide exceptional stability during centrifugation with g-Safe® technology and efficient sample identification with the OptiTrack® Matrix. The design features, including RecoverMax® well design and raised well rims, ensure reliable sealing and facilitate maximum sample recovery.
  • g-Safe®: exceptional centrifugation stability up to 6000×g
  • OptiTrack® Matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labelling
  • RecoverMax® well design: Optimised well geometry for minimal remaining volume and excellent mixing properties
  • Raised well rims and smooth surface for reliable closing, even with heat sealing
  • Unmatched, transparent polypropylene (PP) for improved sample visibility
  • Polypropylene ensures high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without slip agents, plastocizers or biocides (materials which could have a negative effect on bioassays)
  • Ensured functionality from −86 to +100 °C
Pipette, control and recover your samples with ease using Eppendorf Microplates! Designed with the easy-to-read alphanumeric OptiTrack® matrix label to identify individual wells, speed up the filling process and significantly reduce the risk of pipetting errors. Eppendorf Microplates are made with high-transparency polypropylene ensuring quick and easy location of samples and are also available in Eppendorf LoBind® for when you need maximum sample recovery.
Safe code option provides 3-level coding for safe sample and vessel identification. ECC 200 error correction function enables safe code reading with up to 30% damaged codes.
Well colour: Clear
Material: polypropylene (PP)
Autoclavable: 121 °C, 20 min
Centrifugation stability: 6000×g
Plate colour: white
Specifications for Product Family
Specifiche
  • Catalog No:
  • 737-0166
  • 737-0167
  • 737-0168
  • 737-0254
  • 737-0173
  • 737-0174
  • 737-0175
  • 737-0224
  • 737-0255
  • 737-0225
  • 737-0226
  • 737-0229
  • 737-0230
  • Environmentally Preferable:
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Certificazione ambientale:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Specifiche
Product Family Options
96-well
Informazioni sul prodotto
  • N. di pozzetti
    Sterili
    Confezione
    Disponibilità
    Price
  • 737-0166
    Eppendorf® Microplate, flat bottom, PCR clean (Non sterile)
    Eppendorf® Microplate, flat bottom, PCR clean (Non sterile)
    -
    80 plates (5 bags × 16 plates)
    Log in per disponibilità
    Log in per disponibilità
    1 * 80 ST
    1 * 80 ST - SFr. 279.00
    SFr. 279.00
    1 * 80 ST
       SFr. 279.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    737-0166
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    Renewable Energy Certificate, ISO 14001
      Produzione a basso impatto
  • 737-0167
    Eppendorf® Microplate, U-bottom, PCR clean (Non sterile)
    Eppendorf® Microplate, U-bottom, PCR clean (Non sterile)
    96
    -
    80 plates (5 bags × 16 plates)
    Log in per disponibilità
    Log in per disponibilità
    1 * 80 ST
    1 * 80 ST - SFr. 279.00
    SFr. 279.00
    1 * 80 ST
       SFr. 279.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    737-0167
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    Renewable Energy Certificate, ISO 14001
      Produzione a basso impatto
  • 737-0168
    Eppendorf® Microplate, V-bottom, PCR clean (Non sterile)
    Eppendorf® Microplate, V-bottom, PCR clean (Non sterile)
    96
    -
    80 plates (5 bags × 16 plates)
    Log in per disponibilità
    Log in per disponibilità
    1 * 80 ST
    1 * 80 ST - SFr. 279.00
    SFr. 279.00
    1 * 80 ST
       SFr. 279.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    737-0168
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    Renewable Energy Certificate, ISO 14001
      Produzione a basso impatto
  • 737-0254
    Eppendorf® Microplate, 3-level SafeCode (2D datamatrix, linear barcode, alphanumeric code), V-bottom, PCR clean (Non sterile)
    Eppendorf® Microplate, 3-level SafeCode (2D datamatrix, linear barcode, alphanumeric code), V-bottom, PCR clean (Non sterile)
    -
    80 plates (5 bags × 16 plates)
    Log in per disponibilità
    Log in per disponibilità
    1 * 80 ST
    1 * 80 ST - SFr. 422.00
    SFr. 422.00
    1 * 80 ST
       SFr. 422.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    737-0254
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    Renewable Energy Certificate, ISO 14001
      Produzione a basso impatto
  • 737-0173
    Eppendorf® Microplate, flat bottom
    Eppendorf® Microplate, flat bottom
    96
    +
    80 plates (5 bags × 16 plates)
    Log in per disponibilità
    Log in per disponibilità
    1 * 80 ST
    1 * 80 ST - SFr. 320.00
    SFr. 320.00
    1 * 80 ST
       SFr. 320.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    737-0173
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    Renewable Energy Certificate, ISO 14001
      Produzione a basso impatto
  • 737-0174
    Eppendorf® Microplate, U-bottom
    Eppendorf® Microplate, U-bottom
    96
    +
    80 plates (5 bags × 16 plates)
    Log in per disponibilità
    Log in per disponibilità
    1 * 80 ST
    1 * 80 ST - SFr. 320.00
    SFr. 320.00
    1 * 80 ST
       SFr. 320.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    737-0174
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    Renewable Energy Certificate, ISO 14001
      Produzione a basso impatto
  • 737-0175
    Eppendorf® Microplate, V-bottom
    Eppendorf® Microplate, V-bottom
    96
    +
    80 plates (5 bags × 16 plates)
    Log in per disponibilità
    Log in per disponibilità
    1 * 80 ST
    1 * 80 ST - SFr. 320.00
    SFr. 320.00
    1 * 80 ST
       SFr. 320.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    737-0175
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    Renewable Energy Certificate, ISO 14001
      Produzione a basso impatto
  • 737-0224
    Eppendorf® Microplate, V-bottom
    Eppendorf® Microplate, V-bottom
    +
    240 plates (10 bags × 24 plates)
    Log in per disponibilità
    Log in per disponibilità
    1 * 240 ST
    1 * 240 ST - SFr. 861.00
    SFr. 861.00
    1 * 240 ST
       SFr. 861.00
    Specifiche:
    More
    Specifications
    Cat. No.
    737-0224
    Environmentally Preferable
    Certificazione ambientale
384-well
Informazioni sul prodotto
  • N. di pozzetti
    Sterili
    Confezione
    Disponibilità
    Price
  • 737-0255
    Eppendorf® Microplate, 3-level SafeCode (2D datamatrix, linear barcode, alphanumeric code), V-bottom, PCR clean (Non sterile)
    Eppendorf® Microplate, 3-level SafeCode (2D datamatrix, linear barcode, alphanumeric code), V-bottom, PCR clean (Non sterile)
    -
    80 plates (5 bags × 16 plates)
    Log in per disponibilità
    Log in per disponibilità
    1 * 80 ST
    1 * 80 ST - SFr. 836.00
    SFr. 836.00
    1 * 80 ST
       SFr. 836.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    737-0255
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    Renewable Energy Certificate, ISO 14001
      Produzione a basso impatto
  • 737-0225
    Eppendorf® Microplate, flat bottom, PCR clean (Non sterile)
    Eppendorf® Microplate, flat bottom, PCR clean (Non sterile)
    384
    -
    80 plates (5 bags × 16 plates)
    Log in per disponibilità
    Log in per disponibilità
    1 * 80 ST
    1 * 80 ST - SFr. 710.00
    SFr. 710.00
    1 * 80 ST
       SFr. 710.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    737-0225
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    Renewable Energy Certificate, ISO 14001
      Produzione a basso impatto
  • 737-0226
    Eppendorf® Microplate, V-bottom, PCR clean (Non sterile)
    Eppendorf® Microplate, V-bottom, PCR clean (Non sterile)
    384
    -
    80 plates (5 bags × 16 plates)
    Log in per disponibilità
    Log in per disponibilità
    1 * 80 ST
    1 * 80 ST - SFr. 710.00
    SFr. 710.00
    1 * 80 ST
       SFr. 710.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    737-0226
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    Renewable Energy Certificate, ISO 14001
      Produzione a basso impatto
  • 737-0229
    Eppendorf® Microplate, flat bottom
    Eppendorf® Microplate, flat bottom
    384
    +
    80 plates (5 bags × 16 plates)
    Log in per disponibilità
    Log in per disponibilità
    1 * 80 ST
    1 * 80 ST - SFr. 821.00
    SFr. 821.00
    1 * 80 ST
       SFr. 821.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    737-0229
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    Renewable Energy Certificate, ISO 14001
      Produzione a basso impatto
  • 737-0230
    Eppendorf® Microplate, V-bottom
    Eppendorf® Microplate, V-bottom
    384
    +
    80 plates (5 bags × 16 plates)
    Log in per disponibilità
    Log in per disponibilità
    1 * 80 ST
    1 * 80 ST - SFr. 821.00
    SFr. 821.00
    1 * 80 ST
       SFr. 821.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    737-0230
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    Renewable Energy Certificate, ISO 14001
      Produzione a basso impatto