Order Entry
Switzerland
ContactUsLinkComponent
Rodamina B, Supelco®
Rodamina B, Supelco®
Catalog # 79754-25MG
Supplier:  Merck
CAS Number:  
undefined
Rodamina B, Supelco®
Catalog # 79754-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :479.02 g/mol
Merck Index:13,08266
Melting Pt:199…201 °C
Density:1.31 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:201-383-9
 :GHS05
Storage Temperature:Ambiente
CAS Number:81-88-9
Formula:C₂₈H₃₁ClN₂O₃
MDL Number:MFCD00011931