Rodamina B
Catalog # ROTHT130.1
Supplier: Roth Carl
CAS Number:
Specifications
- Tipo di conf.:Glass bottle for solids
- Conf:25 g
- Seq:3308
Specifications
About this item
:
:479.02 g/mol
Merck Index:13,08266
Melting Pt:199…201 °C
Density:1.31 g/cm³ (20 °C)
EINECS:201-383-9
:GHS05
Storage Temperature:Ambiente
CAS Number:81-88-9
Formula:C₂₈H₃₁ClN₂O₃
MDL Number:MFCD00011931