F(ab')2 Anti-Mouse Polyclonal Antibody (CF® 660C)
Supplier: Biotium
Certificates
About this item
Goat anti-mouse IgG (H+L), F(ab')2 fragment secondary antibody labeled with our superior CF® dyes and other labels.
- Available in 14 bright and photostable CF® dyes, and HRP
- Biotin, APC, and R-PE conjugates also available
- Suitable for western, immunofluorescence, and immunohistology in FFPE tissues
Note: Conjugates of blue fluorescent dyes like CF®350 are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colours.
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: CF® 660C
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse IgG
Specifications
- Catalog No:
- 20918-250UL
- 20918-50UL
- Simbolo dell'antigene:
- IgG (H+L)
- IgG (H+L)
- Tipo anticorpo:
- Secondary
- Secondary
- Clonalità:
- Policlonale
- Policlonale
- Coniugazione:
- CF® 660C
- CF® 660C
- Reattività:
- Mouse
- Mouse
- Ospite:
- Goat
- Goat
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescenza:
- Yes
- Yes
- Citometria a flusso:
- Yes
- Yes
- Frammento anticorpo:
- F(ab')2
- F(ab')2
- Concentrazione:
- 2 mg/ml
- 2 mg/ml
- Assorbimento incrociato:
- Not cross-adsorbed
- Not cross-adsorbed
- Temperatura di conservazione:
- Store at −20 °C, protected from light.
- Store at −20 °C, protected from light.