Order Entry
Switzerland
ContactUsLinkComponent
(+)-Indolactam V ≥95% (by 1H-NMR)
Irritant
(+)-Indolactam V ≥95% (by 1H-NMR)
Catalog # ENZOALX420010C300
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Irritant
(+)-Indolactam V ≥95% (by 1H-NMR)
Catalog # ENZOALX420010C300
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Conf:
    300 µG
  • Seq:
    1382

Specifications

Specification Test Results

  • Purity:
    ≥95% (¹H-NMR)
  • Purity Detail:
    May contain traces of (-)-Indolactam V

Specification Test Results

About this item

 :
 :301.4 g/mol
 :GHS07
Storage Temperature:Refrigerata
CAS Number:90365-56-3
Formula:C₁₇H₂₃N₃O₂