Orders LinkContactUsLinkComponent
3-ATA ≥95%
5 mg
  ENZOALX350273M005
3-ATA ≥95%
  ENZOALX350273M005
 :  129821-08-5
3-ATA ≥95%
5 mg
 :  129821-08-5
SFr. 527.00
1 * 5 mg
Specific CDK4 inhibitor (IC50=3.1µM) inducing growth inhibition of p16-altered tumors.
  • CDK4 inhibitor
White to off-white solid powder.
Soluble in DMSO or dimethyl formamide.
 :  3-Amino-9-thio(10H)-acridone
  •  :129821-08-5
  •  :C₁₃H₁₀N₂S
  •  :226.3 g/mol
  •  :Ambiente
  • Conf:
    5 mg