SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
α-Mannosyl ceramide
α-Mannosyl ceramide ≥95% (by TLC, NMR)
α-Mannosyl ceramide ≥95% (by TLC, NMR)
ENZOALX306031M001
:
ENZO LIFE SCIENCES
:
α-Mannosyl ceramide ≥95% (by TLC, NMR)
ENZOALX306031M001
:
ENZO LIFE SCIENCES
:
Conf:
1 mg
Seq:
2070
Purity:
≥95% (TLC and NMR)
:
:
858.3 g/mol
:
Congelamento
:
C₅₀H₉₉NO₉