Orders LinkContactUsLinkComponent
Actinonin ≥98%
Actinonin ≥98%
5 mg
Codice catalogo ENZOALX260128M005
Nome del fornitore:   Enzo Life Sciences
Actinonin ≥98%
Codice catalogo ENZOALX260128M005
Nome del fornitore:  Enzo Life Sciences
Numero CAS:  13434-13-4
Actinonin ≥98%
5 mg
Numero CAS:  13434-13-4
Price
SFr. 210.00
Log in per disponibilità
1 * 5 mg
Product Details & Documents
Protease inhibitor
Antibiotic. Inhibitor of aminopeptidase M, N and leucine and alanyl aminopeptidase and peptide deformylase (PDF). Inhibits MMP-1 (Ki=300nM), MMP-2 (IC50~90nM at MMP-2=5nM), MMP-3 (Ki=1’700nM), MMP-7 (IC50~3’500nM at MMP-7=19nM), MMP-8 (Ki=160nM), MMP-9 (Ki=330nM), MMP-10 (IC50~500nM at MMP-10=22nM), MMP-12 (IC50~350nM at MMP-12=4nM), and MMP-13 (IC50~100nM  at MMP-13=13nM), but not MMP-14 (IC50>1mM at MMP-14=13.6nM). Shows antitumor activity in vitro and in vivo. Apoptosis inducer.
Synthetic.
  • Numero CAS:13434-13-4
Specifiche
Specifiche
  • Conf:
    5 mg