Tablet reagents for Chlorine Combined and Total (Extended Range DPD XF and XT)
PALIPM033
:
- Conf:50
- Descrizione:Tablet reagent for chlorine – free, combined and total (extended range DPD XF and XT)
- Range di misura:0 - 10 mg/L Cl₂
- Test per kit:50
Tablet reagents for chlorine – free, combined and total (extended range DPD XF and XT).
: Starter Packs (PM codes) contain 50 tests plus any optional reagents or equipment needed.