Orders LinkContactUsLinkComponent
EasyFrit ES1210 EasySampler Filter Kit
Supplier:   Mettler - Toledo
EasyFrit ES1210 EasySampler Filter Kit
Supplier:  Mettler - Toledo
Certificates
EasyFrit ES1210 EasySampler Filter Kit
2 Options Available Below
Product Details & Documents
EasyFrit turns EasySampler into a selective solution phase sampling tool for data-rich collection throughout a controlled crystallisation.
  • HPLC ready samples
  • Eliminate user safety concerns
  • Avoid surface fouling
  • Transform and increase productivity
  • Never miss a reaction event
  • Representative and reproducible
  • Material: FEP, alloy C-276, C22 alloy
A unique self-flushing mechanism prevents surface fouling and allows unattended sampling over an elongated period under reaction conditions at scale.
Unattended, representative sampling: EasyFrit is a dynamically flushed in situ filter. Its unique mechanism minimizes the risk of surface fouling. It is compatible with all EasySampler 1210 probes.
Automated and unattended sampling: EasyFrit allows for 24/7 data-rich collection of solution-phase information throughout a controlled crystallization, preventing data blind spots.
Representative and reproducible: EasySampler samples from the same position into a fixed volume pocket for accurate analytical data.
Reactions difficult to sample: Easily manage reactions with high solid loading using automated filtration, sample capture, and dilution.
 : The kit contains one assembled filter and the installation tool.
Product Family Options
Product Information
  • Descrizione
    Materiale
    Ø
    Lunghezza
    Availability
    Price
  • Filter for ES1210 assembled
    — mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Filter kit for ES1210
    FEP
    14,2 mm
    47 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Accessories
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • METB30247094
    Tube set probe 210, PTFE tube, PEEK fittings; 4 pcs in a set