Optical DO Sensor Cable
Optical DO sensor cable
Codice catalogo DISI2700-0418
Nome del fornitore: DISTEK
Vai a
Product Details & Documents
Optical DO sensor cable that will work only with the optical DO sensors such as the Hamilton VisiFerm ECS optical DO sensor.
Specifiche
- Descrizione:Optical DO sensor cable