Orders LinkContactUsLinkComponent
Screw Cap Microtubes, with Cap
Screw Cap Microtubes, with Cap
Nome del fornitore:   DELTALAB
Screw Cap Microtubes, with Cap
Nome del fornitore:  DELTALAB
Certificates
Screw Cap Microtubes, with Cap
3 varianti
Product Details & Documents
Screw cap microtubes, with cap in medical grade, transparent polypropylene. The cap embodies a non-reactive ethylene-propylene o-ring. Suitable for autoclave, liquid nitrogen (gas) and boiling processes. Perfect for long term storage.
  • Withstand temperatures from –190 ℃
  • Centrifugation resistance: 20000×g
  • DNAse, RNAse, DNA, and PCR inhibitors free
  • Graduated models feature a frosted area for writing
  • The non-graduated model incorporates an external grip for an easy handling.
Confezione: Microtubes are supplied capped, in bags of 50 units.
Product Family Options
Informazioni sul prodotto
  • Capacità
    Colore
    Materiale
    Sterili
    Disponibilità
    Price
  • DELT409115/2
    Non skirted microtube with graduation
    Non skirted microtube with graduation
    1,5 ml
    trasparente
    PP
    +
    Log in per disponibilità
    Log in per disponibilità
    1 * 2.500 ST
    1 * 2.500 ST - SFr. 1’850.00
    SFr. 1’850.00
    1 * 2.500 ST
       SFr. 1’850.00
  • DELT409115/3
    Non skirted microtube with graduation
    Non skirted microtube with graduation
    2 ml
    trasparente
    PP
    +
    Log in per disponibilità
    Log in per disponibilità
    1 * 2.500 ST
    1 * 2.500 ST - SFr. 1’800.00
    SFr. 1’800.00
    1 * 2.500 ST
       SFr. 1’800.00
  • DELT409115/6
    Skirted microtube with graduation
    Skirted microtube with graduation
    2 ml
    trasparente
    PP
    +
    Log in per disponibilità
    Log in per disponibilità
    1 * 2.500 ST
    1 * 2.500 ST - SFr. 1’760.00
    SFr. 1’760.00
    1 * 2.500 ST
       SFr. 1’760.00