Neptune® Specialty-Use Pipette Tips
Codice catalogo BITX2342.S
Nome del fornitore: BIOTIX
Specifiche
- Confezione:10 rack da 96
- Sterili:+
- Volume:10 µl
- Seq:02
- Stile punta:Extended length tips
- Colore puntale:Clear
- Conf:960
Specifiche
Informazioni su questo articolo
These precision universal pipette tips are made from virgin polypropylene.
- Autoclavable
- Good sustainability
- Superior stability, firm placement
- Maximum space saving design
Eco-friendly empty pipette tips trays are manufactured from recycled polypropylene. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%.
The extended length facilitates access to deep vessels. The thin wall design ensures an optimal seal and easy ejection with industry leading pipettors. Eco-friendly racks are made from recycled polypropylene and utilize a unique compact packaging configuration that reduces the carbon footprint.
Certifications: Certified free of RNase, DNase, pyrogens, and PCR Inhibitors. Manufactured under stringent controls in an ISO 9001 certified facility. Each and every lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled.