VWR® Wire-O Laboratory Notebooks, Grid Format
Nome del fornitore: VWR Collection
Vai a
Informazioni su questo articolo
Wire-O laboratory notebooks feature a translux cover and grid format pages.
- Translux cover
- 215,9×279,4 mm pages
- 50 pages
- Grid format
Certifications: All books made in accordance with GLP, GMP, and GCP guidelines.
5 varianti
Product Details & Documents
Wire-O laboratory notebooks feature a translux cover and grid format pages.
- Translux cover
- 215,9×279,4 mm pages
- 50 pages
- Grid format
Certifications: All books made in accordance with GLP, GMP, and GCP guidelines.
Product Family Options
Informazioni sul prodotto
- ColoreTipoVersioneFormato cartaNumero di pagineDisponibilitàPrice
- 818-0303Wire-O lab bookWire-O lab booktraslucidoGrid formatEnglish215,9×279,4 mm50Log in per disponibilità1 * 1 STSFr. 57.40
- 818-0312Wire-O lab bookWire-O lab booktraslucidoGrid formatEnglish-German215,9×279,4 mm50Log in per disponibilità1 * 1 STSFr. 57.40
- 818-0313Wire-O lab bookWire-O lab booktraslucidoGrid formatEnglish-French215,9×279,4 mm50Log in per disponibilità1 * 1 STSFr. 57.40
- 818-0314Wire-O lab bookWire-O lab booktraslucidoGrid formatEnglish-Italian215,9×279,4 mm50Log in per disponibilità1 * 1 STSFr. 57.40
- 818-0315Wire-O lab bookWire-O lab booktraslucidoGrid formatEnglish-Spanish215,9×279,4 mm50Log in per disponibilità1 * 1 STSFr. 57.40