Hei-PLATE Sensor Advanced Package Core Magnetic Stirrers
Catalog # HEID506-11157-00
Supplier: Heidolph Instruments GmbH & Co.KG
Specifications
- Descrizione:Hei-PLATE sensor advanced package core
- Conf:1
Specifications
About this item
Hei-PLATE Sensor Advanced Package Core.
- Includes:
- The Hei-PLATE Mix 'n' Heat Core magnetic stirrer (145 mm diameter) - fast and precise heating - without overshoot - for sample-friendly work
- EKT Hei-Con electronic contact thermometer incl. holder
- silicone protective cover for longer lasting protection of your magnetic stirrer