Microflex® Mega Texture 93-256 Disposable Nitrile Gloves
About this item
Microflex® Mega Texture 93-256 Disposable nitrile gloves, provide confident grip and durable protection.
- Orange short cuff nitrile disposable glove with raised grip
Enhanced grip: Featuring our Mega Texture surface for consistent grip in wet and dry conditions
Tear resistance: Robust 0,13 mm/5,1 mil palm thickness to resist tearing in use
High visibility: Orange colour to increase visibility while working
Durable: Strong formulation providing durable yet comfortable hand protection
Food approved: Ideal for a wide range of food handling applications
Recommended for: General purpose auto aftermarket, food processing and handling, oil, fluids and filter change, engine transmission and repair, mechanical jobs, maintenance, tire rotation and changes, vehicle maintenance, assembly of parts.
Certifications: Arrette du 05/08/2020, EC 2023/2006, ASTM D6319, EN ISO 21420:2020, FDA21 CFR 177-2600-US Food contact approved, ISO 13485, ISO 9001, ISO 18889.
Packaging: 100 gloves per dispenser, 10 dispensers per case, 1000 gloves per case.
5 Options Available Below
Product Details & Documents
Microflex® Mega Texture 93-256 Disposable nitrile gloves, provide confident grip and durable protection.
- Orange short cuff nitrile disposable glove with raised grip
Enhanced grip: Featuring our Mega Texture surface for consistent grip in wet and dry conditions
Tear resistance: Robust 0,13 mm/5,1 mil palm thickness to resist tearing in use
High visibility: Orange colour to increase visibility while working
Durable: Strong formulation providing durable yet comfortable hand protection
Food approved: Ideal for a wide range of food handling applications
Recommended for: General purpose auto aftermarket, food processing and handling, oil, fluids and filter change, engine transmission and repair, mechanical jobs, maintenance, tire rotation and changes, vehicle maintenance, assembly of parts.
Certifications: Arrette du 05/08/2020, EC 2023/2006, ASTM D6319, EN ISO 21420:2020, FDA21 CFR 177-2600-US Food contact approved, ISO 13485, ISO 9001, ISO 18889.
Packaging: 100 gloves per dispenser, 10 dispensers per case, 1000 gloves per case.
Specifications for Product Family
Specifications
- Catalog No:
- ANSE93-256/6.5-7
- ANSE93-256/7.5-8
- ANSE93-256/8.5-9
- ANSE93-256/9.5-10
- ANSE93-256/10.5-11
- Modello:
- Mega texture 93-256
- Mega texture 93-256
- Mega texture 93-256
- Mega texture 93-256
- Mega texture 93-256
- Monouso/riutilizzabile:
- monouso
- monouso
- monouso
- monouso
- monouso
- Talcati/non talcati:
- Non talcati
- Non talcati
- Non talcati
- Non talcati
- Non talcati
- Materiale:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Finitura:
- Fully textured
- Fully textured
- Fully textured
- Fully textured
- Fully textured
- Tipo polsino:
- Elastico
- Elastico
- Elastico
- Elastico
- Elastico
- Lunghezza polsino:
- Standard
- Standard
- Standard
- Standard
- Standard
- Spessore delle dita:
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- Spessore palmo:
- 0,13 mm
- 0,13 mm
- 0,13 mm
- 0,13 mm
- 0,13 mm
- Resistenza tessile:
- ≥14 MPa
- ≥14 MPa
- ≥14 MPa
- ≥14 MPa
- ≥14 MPa
- Sterilità:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Antistatico:
- +
- +
- +
- +
- +
- ASTM n.:
- D6319
- D6319
- D6319
- D6319
- D6319
Specifications
Product Family Options
Product Information
- ModelloDimensione del guantoColoreLunghezzaSpessoreAvailabilityPrice
- Mega texture 93-256Sarancione240 mmPalm thickness: 0,13 mm
Finger thickness: 0,16 mmLog in for availability1 * 100 STSFr. 45.60Specifications:More
Specifications
Cat. No.ANSE93-256/6.5-7ModelloMega texture 93-256Monouso/riutilizzabilemonousoTalcati/non talcatiNon talcatiMaterialeNitrileFinituraFully texturedTipo polsinoElasticoLunghezza polsinoStandardSpessore delle dita0,16 mmSpessore palmo0,13 mmResistenza tessile≥14 MPaSterilitàNon sterileAntistatico+ASTM n.D6319 - Mega texture 93-256Marancione240 mmPalm thickness: 0,13 mm
Finger thickness: 0,16 mmLog in for availability1 * 100 STSFr. 45.60Specifications:More
Specifications
Cat. No.ANSE93-256/7.5-8ModelloMega texture 93-256Monouso/riutilizzabilemonousoTalcati/non talcatiNon talcatiMaterialeNitrileFinituraFully texturedTipo polsinoElasticoLunghezza polsinoStandardSpessore delle dita0,16 mmSpessore palmo0,13 mmResistenza tessile≥14 MPaSterilitàNon sterileAntistatico+ASTM n.D6319 - Mega texture 93-256Larancione240 mmPalm thickness: 0,13 mm
Finger thickness: 0,16 mmLog in for availability1 * 100 STSFr. 45.60Specifications:More
Specifications
Cat. No.ANSE93-256/8.5-9ModelloMega texture 93-256Monouso/riutilizzabilemonousoTalcati/non talcatiNon talcatiMaterialeNitrileFinituraFully texturedTipo polsinoElasticoLunghezza polsinoStandardSpessore delle dita0,16 mmSpessore palmo0,13 mmResistenza tessile≥14 MPaSterilitàNon sterileAntistatico+ASTM n.D6319 - Mega texture 93-256XLarancione240 mmPalm thickness: 0,13 mm
Finger thickness: 0,16 mmLog in for availability1 * 100 STSFr. 45.60Specifications:More
Specifications
Cat. No.ANSE93-256/9.5-10ModelloMega texture 93-256Monouso/riutilizzabilemonousoTalcati/non talcatiNon talcatiMaterialeNitrileFinituraFully texturedTipo polsinoElasticoLunghezza polsinoStandardSpessore delle dita0,16 mmSpessore palmo0,13 mmResistenza tessile≥14 MPaSterilitàNon sterileAntistatico+ASTM n.D6319 - Mega texture 93-256XXLarancione240 mmPalm thickness: 0,13 mm
Finger thickness: 0,16 mmLog in for availability1 * 100 STSFr. 45.60Specifications:More
Specifications
Cat. No.ANSE93-256/10.5-11ModelloMega texture 93-256Monouso/riutilizzabilemonousoTalcati/non talcatiNon talcatiMaterialeNitrileFinituraFully texturedTipo polsinoElasticoLunghezza polsinoStandardSpessore delle dita0,16 mmSpessore palmo0,13 mmResistenza tessile≥14 MPaSterilitàNon sterileAntistatico+ASTM n.D6319