CPC® NSH Quick Fittings, ³/₈" Hose Barb Valved In-Line Couplings
Supplier: CPC COLDER
Certificates
About this item
NSH Series couplings are general purpose versions of our pressure-balanced, non-spill, ChemQuik® CQG06 couplings. Moulded polypropylene, EPDM seals and a 100% springless and metal-free flow path provide broad chemical resistance and exceptionally high flow capacity, allowing instant disconnects (and reconnects), even under pressure.
- Non-spill design - Ultimate protection from chemicals and fumes
- Pressure-balanced design - Failsafe disconnect, even under pressure; easy to reconnect at high pressure
- Springless flow path design - Eliminates source of metallic contaminants
- Optional colour coding - Helps prevent accidental misconnects
Their non-spill design virtually eliminates spills and minimizes downtime in a very low cost package.
Spillage: ~0,03 cc per connect
Lubricants: Krytox® PFPE (Inert)
Thumb latch: Glass-filled polypropylene
O-rings: EPDM
Springs (non-wetted): 316 stainless steel
Compression nuts: White polypropylene
Specifications
- Catalog No:
- CPCCNSHD22006
- CPCCNSHD17006
- Pressione di esercizio massima:
- Vacuum to 120 psi, 8,3 bar
- Vacuum to 120 psi, 8,3 bar
- Temperatura d'uso:
- 0...49 °C
- 0...49 °C