Analog Conductivity Measuring Cells for Wastewater
Nome del fornitore: WTW
Certificati
Informazioni su questo articolo
Analog conductivity measuring cells for wastewater are to be connected to analog transmitters.
- Conductivity measuring cell with integrated temperature sensor (up to 100 °C)
- Measuring range 1 µS/cm to 2 S/cm
- Pressure-resistant to 10 bar
- 12,7 mm NPT screw thread
- Shaft: Stainless steel 1,4571
- Cable gland: Brass, nickel-plated
All analog conductivity measuring cells are equipped with an integrated temperature compensation and cover all applications:
In wastewater treatment plants, as flow measurement or in the pipeline: The right solution for every application
Highest linearity by four electrode measuring cell
High operational safety by robust design
Electrical connection: Integrated PU connection cable with 7-pole screw plug (IP 65).
Specifiche
- Catalog No:
- WTWA302222
- WTWA302220
- WTWA302225
- Tipo di sensore:
- Integrated NTC
- Integrated NTC
- Integrated NTC
- Intervallo di conducibilità:
- 0,001...200 µS/cm
- 0,001...200 µS/cm
- 1 µS/cm...2 S/cm
- Costante di cella:
- 0,1 cm⁻¹, ±2%
- 0,1 cm⁻¹, ±2%
- 0,475 cm⁻¹, ±1,5%